Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004840.hg.1 |
ENST00000241014.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004840.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004840.hg.1 |
ENST00000478807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004840.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004840.hg.1 |
TC11000389.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004839.hg.1 |
ENST00000241014.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004839.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004839.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004839.hg.1 |
TC11000389.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004841.hg.1 |
ENST00000241014.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004841.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004841.hg.1 |
ENST00000478807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004841.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004841.hg.1 |
TC11000389.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152581.hg.1 |
TC1100007429.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
ENST00000241014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
ENST00000395629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
NM_005456.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152580.hg.1 |
TC1100007429.hg.1 |
Clariom D TC |