Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000580896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000580896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000582786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000582786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000582786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023644.hg.1 |
ENST00000262436.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023644.hg.1 |
ENST00000397786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023644.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023644.hg.1 |
TC17001772.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006653.hg.1 |
ENST00000428342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006653.hg.1 |
ENST00000428342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000578132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000580896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000580896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000581994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000582786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000582786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000582786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011369.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
ENST00000397786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
ENST00000583958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
NM_005121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700191520.hg.1 |
TC1700011369.hg.1 |
Clariom D TC |