Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004788.3 | NP_004779.2 | 6 - 7 | 878 | 71 | |
GenBank mRNA | D50916.1 | - | 6 - 7 | 815 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |