Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001447.2 | NP_001438.1 | 16 - 17 | 10325 | 68 | |
XM_006714761.3 | XP_006714824.1 | 17 - 18 | 10896 | 68 | ||
XM_011537600.2 | XP_011535902.1 | 19 - 20 | 10664 | 68 | ||
XM_011537603.2 | XP_011535905.1 | 17 - 18 | 10597 | 68 | ||
XM_017009224.1 | XP_016864713.1 | 19 - 20 | 10666 | 68 | ||
XM_017009225.1 | XP_016864714.1 | 18 - 19 | 10508 | 68 | ||
GenBank mRNA | AB160984.1 | - | 4 - 5 | 893 | 68 | |
AF231022.1 | - | 16 - 17 | 10325 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |