Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000474309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000474309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000474309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020006.hg.1 |
ENST00000370884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020006.hg.1 |
ENST00000370886.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020006.hg.1 |
ENST00000370902.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020006.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020006.hg.1 |
TC10001566.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178622.hg.1 |
TC1000011552.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370885.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370886.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000370902.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000471049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000472345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000474309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000474309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000474309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000476234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000477692.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000485122.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
ENST00000489158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011552.hg.1 |
NM_016046.4 |
Refseq |