Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015167.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
ENST00000412586.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
ENST00000423703.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
ENST00000444704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015171.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
ENST00000412586.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
ENST00000423703.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015148.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
ENST00000412586.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
ENST00000423703.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
ENST00000444704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015168.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015175.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015170.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015176.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015169.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015172.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
ENST00000341550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
ENST00000396807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
ENST00000446105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
NM_001127585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015174.hg.1 |
TC12001146.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000341550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000396807.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000412586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000423703.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000437149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000446105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000467678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000469749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000472002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000479301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000482489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000488381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000493267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000493873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000619641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000341550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000396807.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000412586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000423703.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000437149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000446105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000467678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000469749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000472002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000479301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000482489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000488381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000493267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000493873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
ENST00000619641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127582.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_001127586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
NM_016162.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173006.hg.1 |
TC1200009747.hg.1 |
Clariom D TC |