Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016943.hg.1 |
TC09001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016943.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016943.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016941.hg.1 |
TC09001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016941.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016941.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016942.hg.1 |
TC09001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016942.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016942.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016944.hg.1 |
TC09001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016944.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016944.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173858.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173858.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173858.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173858.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173858.hg.1 |
TC0900010865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173856.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173856.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173856.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173856.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173856.hg.1 |
TC0900010865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173860.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173860.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173860.hg.1 |
TC0900010865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173857.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173857.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173857.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173857.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173857.hg.1 |
TC0900010865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173859.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173859.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173859.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173859.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173859.hg.1 |
TC0900010865.hg.1 |
Clariom D TC |