Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015037.hg.1 |
TC03000851.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015037.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015037.hg.1 |
ENST00000496455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015037.hg.1 |
ENST00000537975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015037.hg.1 |
ENST00000541628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015037.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000476145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009271.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
ENST00000295920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
ENST00000496455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
NM_003875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166435.hg.1 |
TC0300009271.hg.1 |
Clariom D TC |