Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015190.hg.1 |
ENST00000303383.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015190.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015190.hg.1 |
TC16001083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000564272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000564272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178304.hg.1 |
TC1600010168.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000564272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000564272.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000565887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000566016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000567698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010168.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
ENST00000303383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
ENST00000563219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
ENST00000569702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
NM_024745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178303.hg.1 |
TC1600010168.hg.1 |
Clariom D TC |