Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005751.4 | NP_005742.4 | 22 - 23 | 5828 | 96 | |
NM_147185.2 | NP_671714.1 | 22 - 23 | 5828 | 96 | ||
XM_017011642.1 | XP_016867131.1 | 23 - 24 | 5932 | 96 | ||
XM_017011643.1 | XP_016867132.1 | 23 - 24 | 5932 | 96 | ||
XM_017011644.1 | XP_016867133.1 | 23 - 24 | 5932 | 96 | ||
XM_017011645.1 | XP_016867134.1 | 22 - 23 | 5878 | 96 | ||
XM_017011646.1 | XP_016867135.1 | 23 - 24 | 5932 | 96 | ||
XM_017011647.1 | XP_016867136.1 | 22 - 23 | 5839 | 96 | ||
XM_017011648.1 | XP_016867137.1 | 22 - 23 | 5836 | 96 | ||
XM_017011649.1 | XP_016867138.1 | 23 - 24 | 5932 | 96 | ||
XM_017011650.1 | XP_016867139.1 | 22 - 23 | 5836 | 96 | ||
XM_017011651.1 | XP_016867140.1 | 22 - 23 | 5791 | 96 | ||
XM_017011652.1 | XP_016867141.1 | 23 - 24 | 5932 | 96 | ||
XM_017011653.1 | XP_016867142.1 | 21 - 22 | 5743 | 96 | ||
XM_017011654.1 | XP_016867143.1 | 21 - 22 | 5695 | 96 | ||
XM_017011655.1 | XP_016867144.1 | 23 - 24 | 5591 | 96 | ||
XM_017011656.1 | XP_016867145.1 | 22 - 23 | 5521 | 96 | ||
XM_017011657.1 | XP_016867146.1 | 8 - 9 | 1441 | 96 | ||
GenBank mRNA | AF083037.1 | - | 15 - 16 | 4667 | 96 | |
AF247727.4 | - | 21 - 22 | 5462 | 96 | ||
AJ131693.1 | - | 22 - 23 | 5828 | 96 | ||
AY528715.1 | - | 4 - 5 | 675 | 96 | ||
BC062448.1 | - | 1 - 2 | 199 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |