Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029182.hg.1 |
TC03001613.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029182.hg.1 |
ENST00000193391.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029182.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188135.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188135.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188135.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188135.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188135.hg.1 |
TC0300011852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
ENST00000193391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011852.hg.1 |
NM_016247.3 |
Refseq |