Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020796.4 | NP_065847.1 | 16 - 17 | 2495 | 98 | |
GenBank mRNA | AF279656.1 | - | 16 - 17 | 2372 | 98 | |
AK027471.1 | - | 4 - 5 | 448 | 98 | ||
AK027501.1 | - | 7 - 8 | 806 | 98 | ||
AK027654.1 | - | 4 - 5 | 455 | 98 | ||
AK074975.1 | - | 4 - 5 | 366 | 98 | ||
AK315143.1 | - | 16 - 17 | 1914 | 98 | ||
BC032619.1 | - | 16 - 17 | 2506 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |