Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010044.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010044.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010044.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010044.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010044.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174370.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400174370.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174370.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400174370.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174370.hg.1 |
TC1400010044.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010044.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010044.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010044.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010044.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010044.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174372.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400174372.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174372.hg.1 |
ENST00000553483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400174372.hg.1 |
NR_110563.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174372.hg.1 |
TC1400010044.hg.1 |
Clariom D TC |