Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014326.hg.1 |
TC0X001063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014326.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014326.hg.1 |
ENST00000375368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014326.hg.1 |
ENST00000396435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014326.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014326.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166637.hg.1 |
TC0X00009755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |