Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11008521.hg.1 |
ENST00000502692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11008521.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11008521.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11008521.hg.1 |
TC11000671.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100159181.hg.1 |
TC1100008090.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000502692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000511191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
ENST00000513398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001104.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008090.hg.1 |
NM_001258371.2 |
Refseq |