Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004564.hg.1 |
ENST00000359358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004564.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004564.hg.1 |
TC18000447.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004563.hg.1 |
ENST00000359358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004563.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004563.hg.1 |
TC18000447.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000583263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000583263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
ENST00000583263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000358095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000583263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000583263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
ENST00000583263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008427.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157387.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157387.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157387.hg.1 |
TC1800008427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157387.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157387.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157388.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157388.hg.1 |
NM_020805.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157388.hg.1 |
TC1800008427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157388.hg.1 |
ENST00000359358.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157388.hg.1 |
NM_020805.2 |
Refseq |