Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291476.1 | NP_001278405.1 | 14 - 15 | 1806 | 96 | |
NM_007010.4 | NP_008941.3 | 13 - 14 | 1691 | 96 | ||
XM_011524232.2 | XP_011522534.1 | 13 - 14 | 1675 | 96 | ||
XM_011524233.2 | XP_011522535.1 | 11 - 12 | 1547 | 96 | ||
GenBank mRNA | AB208938.1 | - | 11 - 12 | 2856 | 96 | |
AF077033.1 | - | 13 - 14 | 1658 | 96 | ||
AJ010840.1 | - | 10 - 11 | 1110 | 96 | ||
AK001245.1 | - | 10 - 11 | 1200 | 96 | ||
AK001652.1 | - | 14 - 15 | 1787 | 96 | ||
AK025721.1 | - | 3 - 4 | 193 | 96 | ||
AK027425.1 | - | 10 - 11 | 1151 | 96 | ||
BC041785.1 | - | 13 - 14 | 1673 | 4808288 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |