Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004605.hg.1 |
TC03003346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004605.hg.1 |
ENST00000383748.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004605.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004605.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013815.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
ENST00000383748.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
ENST00000418176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
ENST00000463888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
NM_001205272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203871.hg.1 |
TC0300013815.hg.1 |
Clariom D TC |