Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015132.4 | NP_055947.1 | 16 - 17 | 1848 | 86 | |
XM_005249672.1 | XP_005249729.1 | 16 - 17 | 1858 | 86 | ||
XM_011515229.1 | XP_011513531.1 | 16 - 17 | 1858 | 86 | ||
XM_011515230.2 | XP_011513532.1 | 17 - 18 | 1905 | 86 | ||
XM_011515231.2 | XP_011513533.1 | 18 - 19 | 1964 | 86 | ||
XM_011515233.2 | XP_011513535.1 | 17 - 18 | 1906 | 86 | ||
XM_017011893.1 | XP_016867382.1 | 16 - 17 | 1858 | 86 | ||
XM_017011894.1 | XP_016867383.1 | 17 - 18 | 2305 | 86 | ||
XM_017011895.1 | XP_016867384.1 | 17 - 18 | 1910 | 86 | ||
XM_017011896.1 | XP_016867385.1 | 17 - 18 | 3818 | 86 | ||
XM_017011897.1 | XP_016867386.1 | 16 - 17 | 1779 | 86 | ||
GenBank mRNA | AB018256.1 | - | 16 - 17 | 1808 | 86 | |
AF420470.1 | - | 16 - 17 | 1816 | 86 | ||
AK023740.1 | - | 11 - 12 | 1093 | 86 | ||
AK294582.1 | - | 18 - 19 | 1803 | 86 | ||
AK315135.1 | - | 16 - 17 | 1834 | 86 | ||
BC045667.1 | - | 16 - 17 | 1818 | 5298570 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |