Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014845.5 | NP_055660.1 | 13 - 14 | 1651 | 76 | |
XM_011536281.2 | XP_011534583.1 | 13 - 14 | 2183 | 76 | ||
XM_017011591.1 | XP_016867080.1 | 13 - 14 | 1656 | 76 | ||
XM_017011592.1 | XP_016867081.1 | 12 - 13 | 1419 | 76 | ||
XM_017011593.1 | XP_016867082.1 | 11 - 12 | 1354 | 76 | ||
GenBank mRNA | AK222732.1 | - | 13 - 14 | 1548 | 76 | |
AK291671.1 | - | 13 - 14 | 1576 | 76 | ||
AK293422.1 | - | 8 - 9 | 902 | 76 | ||
BC041338.2 | - | 13 - 14 | 1607 | 5274004 | 76 | |
D87464.1 | - | 13 - 14 | 1560 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |