Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022809.hg.1 |
TC01006290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022809.hg.1 |
ENST00000367821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022809.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177513.hg.1 |
TC0100010526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000367821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000431969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000441464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
ENST00000465440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010526.hg.1 |
NM_005149.2 |
Refseq |