Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010853.hg.1 |
TC09000828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010853.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010853.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010854.hg.1 |
TC09000828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010854.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010854.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163174.hg.1 |
TC0900009247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163173.hg.1 |
TC0900009247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163172.hg.1 |
TC0900009247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |