Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030651.hg.1 |
TC07001995.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030651.hg.1 |
ENST00000286091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030651.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193496.hg.1 |
TC0700012968.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000286091.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000413966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
ENST00000466592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012968.hg.1 |
NM_004911.4 |
Refseq |