Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175209.hg.1 |
TC0600011118.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000265425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000360488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
ENST00000377850.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_001286125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011118.hg.1 |
NM_005835.3 |
Refseq |