Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003663.hg.1 |
TC03000189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003663.hg.1 |
ENST00000311856.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003663.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
ENST00000415844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007073.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149473.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149473.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149473.hg.1 |
ENST00000311856.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149473.hg.1 |
NM_004256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149473.hg.1 |
TC0300007073.hg.1 |
Clariom D TC |