Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286242.1 | NP_001273171.1 | 22 - 23 | 3552 | 71 | |
NM_001520.3 | NP_001511.2 | 22 - 23 | 3552 | 71 | ||
GenBank mRNA | BC044857.2 | - | 2 - 3 | 194 | 71 | |
BC137229.1 | - | 22 - 23 | 3531 | 9021226 | 71 | |
BX648143.1 | - | 22 - 23 | 3521 | 71 | ||
CR627430.1 | - | 22 - 23 | 3537 | 71 | ||
U02619.1 | - | 22 - 23 | 3572 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |