Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017020609.1 | XP_016876098.1 | 1 - 2 | 117 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |