Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011291.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011291.hg.1 |
ENST00000373694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011291.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011291.hg.1 |
TC20000821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011293.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011293.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011293.hg.1 |
TC20000821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011290.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011290.hg.1 |
ENST00000373694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011290.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011290.hg.1 |
TC20000821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011292.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011292.hg.1 |
ENST00000373694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011292.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011292.hg.1 |
TC20000821.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000163220.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163220.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163220.hg.1 |
TC2000009023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163220.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163220.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163219.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163219.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163219.hg.1 |
TC2000009023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163219.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163219.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163221.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163221.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163221.hg.1 |
TC2000009023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163221.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163221.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163218.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163218.hg.1 |
NM_015474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163218.hg.1 |
TC2000009023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163218.hg.1 |
ENST00000262878.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163218.hg.1 |
NM_015474.3 |
Refseq |