Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522547.2 | XP_011520849.1 | 11 - 12 | 1209 | 105 | |
XM_011522548.2 | XP_011520850.1 | 10 - 11 | 1128 | 105 | ||
XM_017023322.1 | XP_016878811.1 | 13 - 14 | 3073 | 105 | ||
XM_017023323.1 | XP_016878812.1 | 11 - 12 | 1230 | 105 | ||
XM_017023326.1 | XP_016878815.1 | 14 - 15 | 2102 | 105 | ||
XM_017023328.1 | XP_016878817.1 | 14 - 15 | 2011 | 105 | ||
XM_017023330.1 | XP_016878819.1 | 13 - 14 | 2550 | 105 | ||
XM_017023332.1 | XP_016878821.1 | 11 - 12 | 1230 | 105 | ||
XM_017023335.1 | XP_016878824.1 | 14 - 15 | 2102 | 105 | ||
XM_017023336.1 | XP_016878825.1 | 13 - 14 | 1112 | 105 | ||
XM_017023337.1 | XP_016878826.1 | 10 - 11 | 1149 | 105 | ||
XM_017023339.1 | XP_016878828.1 | 11 - 12 | 1256 | 105 | ||
XM_017023341.1 | XP_016878830.1 | 13 - 14 | 2021 | 105 | ||
XM_017023342.1 | XP_016878831.1 | 12 - 13 | 1031 | 105 | ||
XM_017023343.1 | XP_016878832.1 | 10 - 11 | 1178 | 105 | ||
GenBank mRNA | AK299664.1 | - | 11 - 12 | 890 | 105 | |
KJ535000.1 | - | 11 - 12 | 1052 | 105 | ||
KJ535001.1 | - | 10 - 11 | 971 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |