Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019969.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019969.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019969.hg.1 |
TC12001521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010753.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181916.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181916.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181916.hg.1 |
ENST00000257974.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181916.hg.1 |
NM_033033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181916.hg.1 |
TC1200010753.hg.1 |
Clariom D TC |