Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291815.1 | NP_001278744.1 | 80 - 81 | 12151 | 94 | |
XM_011518465.2 | XP_011516767.1 | 80 - 81 | 12288 | 94 | ||
XM_011518466.2 | XP_011516768.1 | 80 - 81 | 12288 | 94 | ||
XM_011518467.2 | XP_011516769.1 | 79 - 80 | 12111 | 94 | ||
XM_011518468.2 | XP_011516770.1 | 80 - 81 | 12288 | 94 | ||
XM_017014585.1 | XP_016870074.1 | 59 - 60 | 8957 | 94 | ||
XM_017014586.1 | XP_016870075.1 | 34 - 35 | 4885 | 94 | ||
GenBank mRNA | AK093583.1 | - | 6 - 7 | 734 | 94 | |
AL834139.1 | - | 21 - 22 | 2800 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |