Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005470.hg.1 |
TC01000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005470.hg.1 |
ENST00000374141.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005470.hg.1 |
ENST00000374142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005470.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005470.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005467.hg.1 |
TC01000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005467.hg.1 |
ENST00000374141.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005467.hg.1 |
ENST00000374142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005467.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005468.hg.1 |
TC01000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005468.hg.1 |
ENST00000374141.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005468.hg.1 |
ENST00000374142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005468.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005469.hg.1 |
TC01000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005469.hg.1 |
ENST00000374141.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005469.hg.1 |
ENST00000374142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005469.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152369.hg.1 |
TC0100007484.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000478902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007484.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152367.hg.1 |
TC0100007484.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000488397.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152368.hg.1 |
TC0100007484.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
ENST00000374141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
ENST00000374142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
ENST00000534643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
NM_032283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152366.hg.1 |
TC0100007484.hg.1 |
Clariom D TC |