Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029161.hg.1 |
TC01001874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029161.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029161.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029162.hg.1 |
TC01001874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029162.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029162.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011838.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187294.hg.1 |
TC0100011838.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100187297.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187297.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187297.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187297.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187297.hg.1 |
TC0100011838.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100187296.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187296.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187296.hg.1 |
ENST00000366757.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187296.hg.1 |
NM_183062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187296.hg.1 |
TC0100011838.hg.1 |
Clariom D TC |