Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021881.hg.1 |
TC08001675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021881.hg.1 |
ENST00000303045.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021881.hg.1 |
ENST00000341807.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021881.hg.1 |
ENST00000435777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021881.hg.1 |
ENST00000545577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021881.hg.1 |
NM_152888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
ENST00000303045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
ENST00000341807.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
ENST00000435777.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
NM_152888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
ENST00000303045.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
ENST00000341807.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
ENST00000435777.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
NM_152888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800181246.hg.1 |
TC0800011954.hg.1 |
Clariom D TC |