Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009459.hg.1 |
TC0X000735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009459.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009463.hg.1 |
TC0X000735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009463.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009463.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009462.hg.1 |
TC0X000735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009462.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009462.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009460.hg.1 |
TC0X000735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009460.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159378.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159378.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159378.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159378.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159378.hg.1 |
TC0X00008737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159375.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159375.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159375.hg.1 |
TC0X00008737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159374.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159374.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159374.hg.1 |
TC0X00008737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159377.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159377.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159377.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159377.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159377.hg.1 |
TC0X00008737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159379.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159379.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159379.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159379.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159379.hg.1 |
TC0X00008737.hg.1 |
Clariom D TC |