Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207437.3 | NP_997320.2 | 66 - 67 | 11429 | 139 | |
XM_005253555.3 | XP_005253612.1 | 67 - 68 | 12092 | 139 | ||
XM_011538014.2 | XP_011536316.1 | 66 - 67 | 11933 | 139 | ||
XM_011538015.2 | XP_011536317.1 | 66 - 67 | 11903 | 139 | ||
XM_011538016.2 | XP_011536318.1 | 66 - 67 | 11903 | 139 | ||
XM_011538017.2 | XP_011536319.1 | 65 - 66 | 11756 | 139 | ||
XM_011538019.2 | XP_011536321.1 | 49 - 50 | 8691 | 139 | ||
XM_017018960.1 | XP_016874449.1 | 65 - 66 | 11807 | 139 | ||
XM_017018961.1 | XP_016874450.1 | 66 - 67 | 11783 | 139 | ||
XM_017018962.1 | XP_016874451.1 | 49 - 50 | 8613 | 139 | ||
GenBank mRNA | AB095937.2 | - | 41 - 42 | 7144 | 139 | |
AB290168.1 | - | 46 - 47 | 7948 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |