Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007349.3 | NP_031375.3 | 15 - 16 | 2702 | 70 | |
XM_005249539.1 | XP_005249596.1 | 16 - 17 | 2841 | 70 | ||
XM_011515982.2 | XP_011514284.1 | 15 - 16 | 2816 | 70 | ||
XM_011515983.1 | XP_011514285.1 | 15 - 16 | 2702 | 70 | ||
XM_017011881.1 | XP_016867370.1 | 16 - 17 | 4014 | 70 | ||
GenBank mRNA | AK093584.1 | - | 9 - 10 | 1395 | 70 | |
AK123600.1 | - | 16 - 17 | 2841 | 70 | ||
AK126674.1 | - | 7 - 8 | 2710 | 70 | ||
AK293744.1 | - | 13 - 14 | 2534 | 70 | ||
BC008328.1 | - | 6 - 7 | 626 | 70 | ||
BC033781.1 | - | 10 - 11 | 1943 | 4509264 | 70 | |
BX538201.1 | - | 9 - 10 | 1478 | 70 | ||
BX640616.1 | - | 15 - 16 | 4600 | 70 | ||
U80735.1 | - | 11 - 12 | 1685 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |