Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008272.hg.1 |
TC04000566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008272.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008272.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157676.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400157676.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157676.hg.1 |
ENST00000317735.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400157676.hg.1 |
NM_006583.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157676.hg.1 |
TC0400008389.hg.1 |
Clariom D TC |