Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009777.hg.1 |
ENST00000442948.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009777.hg.1 |
ENST00000445078.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009777.hg.1 |
ENST00000456237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009777.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009777.hg.1 |
TC11000751.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009773.hg.1 |
ENST00000442948.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009773.hg.1 |
ENST00000445078.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009773.hg.1 |
ENST00000456237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009773.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009773.hg.1 |
TC11000751.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000545379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000545379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000545379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202042.hg.1 |
TC1100013069.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202043.hg.1 |
TC1100013069.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000442948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000545379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000545379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000545379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000546166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000611028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
ENST00000612844.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013069.hg.1 |
NM_000804.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000538934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008339.hg.1 |
ENST00000539482.1 |
Ensembl |