Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007831.hg.1 |
TC06000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007831.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007831.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007831.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008229.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
ENST00000008391.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
ENST00000492804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
NM_172238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157542.hg.1 |
TC0600008229.hg.1 |
Clariom D TC |