Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000496008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000496008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000496008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014196.hg.1 |
TC06001185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014196.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014196.hg.1 |
ENST00000354419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014196.hg.1 |
ENST00000443060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014196.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014196.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600169979.hg.1 |
TC0600010241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000351261.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000354419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000443060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000496008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000496008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000496008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000504593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
ENST00000515361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_005355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010241.hg.1 |
NM_030615.2 |
Refseq |