Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278530.1 | NP_001265459.1 | 20 - 21 | 4214 | 70 | |
XM_005267844.2 | XP_005267901.1 | 20 - 21 | 4417 | 70 | ||
XM_017021437.1 | XP_016876926.1 | 16 - 17 | 2674 | 70 | ||
GenBank mRNA | AK001219.1 | - | 5 - 6 | 731 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |