Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
TC02000750.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000354888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000393106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000393108.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000393110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000409811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
ENST00000450943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011661.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
TC02000750.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000354888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000393106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000393108.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000393110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000409811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000425223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
ENST00000450943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011668.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163406.hg.1 |
TC0200009060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163411.hg.1 |
TC0200009060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393106.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393107.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000393110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
ENST00000409811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_001008410.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_018234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009060.hg.1 |
NM_182915.2 |
Refseq |