Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300867.1 | NP_001287796.1 | 14 - 15 | 1516 | 77 | |
NM_001324062.1 | NP_001310991.1 | 19 - 20 | 4718 | 77 | ||
NM_001324063.1 | NP_001310992.1 | 17 - 18 | 4541 | 77 | ||
NM_001324064.1 | NP_001310993.1 | 20 - 21 | 4763 | 77 | ||
NM_001324065.1 | NP_001310994.1 | 14 - 15 | 1516 | 77 | ||
NM_198551.3 | NP_940953.2 | 19 - 20 | 4718 | 77 | ||
XM_006711304.3 | XP_006711367.2 | 17 - 18 | 4853 | 77 | ||
XM_017001243.1 | XP_016856732.1 | 19 - 20 | 4543 | 77 | ||
GenBank mRNA | AK025122.1 | - | 11 - 12 | 1017 | 77 | |
AK025506.1 | - | 16 - 17 | 2398 | 77 | ||
AK290335.1 | - | 10 - 11 | 993 | 77 | ||
AK296712.1 | - | 14 - 15 | 1497 | 77 | ||
AX775845.1 | - | 14 - 15 | 1490 | 77 | ||
BC047116.1 | - | 4 - 5 | 390 | 77 | ||
D87742.1 | - | 16 - 17 | 2567 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |