Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001838.hg.1 |
TC02004916.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001838.hg.1 |
ENST00000344420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001838.hg.1 |
ENST00000416475.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001838.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001838.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001837.hg.1 |
TC02004916.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001837.hg.1 |
ENST00000344420.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001837.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221217.hg.1 |
TC0200016416.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000414029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
ENST00000482746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016416.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
ENST00000344420.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
ENST00000429494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
NM_017877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221216.hg.1 |
TC0200016416.hg.1 |
Clariom D TC |