Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000462854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000462854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000462854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000491170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000491170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000462854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000462854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000462854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000470675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000491170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000491170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016510.hg.1 |
ENST00000497526.1 |
Ensembl |