Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019950.hg.1 |
TC03003327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019950.hg.1 |
ENST00000245046.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019950.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019950.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206546.hg.1 |
TC0300013934.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000245046.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000429759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000487465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
ENST00000497557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013934.hg.1 |
NM_018447.3 |
Refseq |