Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016467.hg.1 |
TC06001439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016467.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016467.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016464.hg.1 |
TC06001439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016464.hg.1 |
ENST00000361028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016464.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016464.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016464.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016465.hg.1 |
TC06001439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016465.hg.1 |
ENST00000361028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016465.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016465.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016465.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175926.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175926.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175926.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175926.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175926.hg.1 |
TC0600011253.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175923.hg.1 |
TC0600011253.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175924.hg.1 |
TC0600011253.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000361028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
ENST00000396827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NM_001163391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011253.hg.1 |
NR_028077.1 |
Refseq |