Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023098.hg.1 |
TC07001536.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023098.hg.1 |
ENST00000222902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023098.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023098.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023099.hg.1 |
TC07001536.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023099.hg.1 |
ENST00000222902.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023099.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023099.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011561.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182287.hg.1 |
TC0700011561.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182288.hg.1 |
TC0700011561.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000222902.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
ENST00000416943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
NM_002991.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011561.hg.1 |
NM_002991.2 |
Refseq |