Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000308.3 | NP_000299.2 | 10 - 11 | 2182 | 63 | |
NM_001127695.2 | NP_001121167.1 | 10 - 11 | 2228 | 63 | ||
NM_001167594.2 | NP_001161066.1 | 9 - 10 | 2131 | 63 | ||
NR_133656.1 | 10 - 11 | 2189 | 63 | |||
GenBank mRNA | AB209705.2 | - | 10 - 11 | 998 | 63 | |
AK097786.1 | - | 9 - 10 | 895 | 63 | ||
AK172808.1 | - | 10 - 11 | 2186 | 63 | ||
AK307701.1 | - | 10 - 11 | 971 | 63 | ||
AK312898.1 | - | 10 - 11 | 991 | 63 | ||
BC000597.2 | - | 10 - 11 | 984 | 3346150 | 63 | |
BC093009.1 | - | 10 - 11 | 991 | 30348898 | 63 | |
M22960.1 | - | 9 - 10 | 952 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |